Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100745.2 | NP_563824.1 | 12 - 13 | 2870 | 78 | |
GenBank mRNA | AF305913.1 | - | 12 - 13 | 2781 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |