Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331786.1 | NP_001322729.1 | 6 - 7 | 2291 | 83 | |
NM_100745.2 | NP_563824.1 | 6 - 7 | 2291 | 83 | ||
GenBank mRNA | AF305913.1 | - | 6 - 7 | 2202 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |