Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XIC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331787.1 | NP_001321098.1 | 2 - 3 | 662 | 75 | |
NM_100746.3 | NP_172349.2 | 2 - 3 | 161 | 75 | ||
GenBank mRNA | LC177512.1 | - | 1 - 2 | 137 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |