Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VAP27-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035927.2 | NP_001031004.1 | 7 - 8 | 1349 | 73 | |
NM_001331791.1 | NP_001321657.1 | 7 - 8 | 1942 | 73 | ||
NM_001331792.1 | NP_001321655.1 | 7 - 8 | 1788 | 73 | ||
NM_001331793.1 | NP_001321656.1 | 7 - 8 | 1724 | 73 | ||
NM_100756.2 | NP_172359.2 | 8 - 9 | 1267 | 73 | ||
GenBank mRNA | AK317489.1 | - | 7 - 8 | 1122 | 73 | |
AK317626.1 | - | 8 - 9 | 1268 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |