Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BON3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100759.4 | NP_172362.3 | 3 - 4 | 540 | 147 | |
GenBank mRNA | AY741136.1 | - | 3 - 4 | 356 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |