Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035929.1 | NP_001031006.1 | 8 - 9 | 852 | 97 | |
NM_100765.5 | NP_563830.1 | 8 - 9 | 1017 | 97 | ||
GenBank mRNA | AJ249967.1 | - | 8 - 9 | 809 | 97 | |
BT008661.1 | - | 8 - 9 | 827 | 97 | ||
D89051.1 | - | 8 - 9 | 752 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |