Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100769.4 | NP_563831.1 | 5 - 6 | 801 | 139 | |
GenBank mRNA | AF202077.1 | - | 5 - 6 | 721 | 139 | |
AY037198.1 | - | 5 - 6 | 727 | 139 | ||
BT004554.1 | - | 5 - 6 | 697 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |