Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AT1G09010 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100772.5 | NP_563833.2 | 1 - 2 | 309 | 93 | |
GenBank mRNA | AB122060.1 | - | 1 - 2 | 150 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |