Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: G6PD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198018.1 | NP_001184947.1 | 4 - 5 | 804 | 134 | |
NM_100813.2 | NP_563844.1 | 4 - 5 | 750 | 134 | ||
GenBank mRNA | AK229085.1 | - | 4 - 5 | 767 | 134 | |
AY056232.1 | - | 4 - 5 | 735 | 134 | ||
AY117271.1 | - | 4 - 5 | 646 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |