Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: U2A' | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100848.4 | NP_172447.1 | 3 - 4 | 345 | 139 | |
GenBank mRNA | AY059902.1 | - | 3 - 4 | 308 | 139 | |
BT000143.1 | - | 3 - 4 | 256 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |