Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COBL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331851.1 | NP_001322459.1 | 5 - 6 | 1229 | 130 | |
NM_100851.2 | NP_172450.2 | 5 - 6 | 1063 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |