Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNL6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331906.1 | NP_001321764.1 | 7 - 8 | 1080 | 92 | |
NM_001331907.1 | NP_001321765.1 | 6 - 7 | 867 | 92 | ||
NM_100919.5 | NP_172515.2 | 7 - 8 | 1080 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |