Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331909.1 | NP_001321924.1 | 4 - 5 | 453 | 78 | |
NM_100921.3 | NP_172517.1 | 3 - 4 | 938 | 78 | ||
GenBank mRNA | AB008487.1 | - | 3 - 4 | 453 | 78 | |
AB010915.1 | - | 3 - 4 | 535 | 78 | ||
AF057282.1 | - | 3 - 4 | 412 | 78 | ||
AK226322.1 | - | 3 - 4 | 397 | 78 | ||
BT024478.1 | - | 3 - 4 | 337 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |