Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XTH33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100930.4 | NP_172525.1 | 1 - 2 | 300 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |