Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGP10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331921.1 | NP_001320481.1 | 5 - 6 | 945 | 76 | |
NM_100944.2 | NP_172538.1 | 5 - 6 | 1088 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |