Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331926.1 | NP_001318975.1 | 24 - 25 | 3290 | 74 | |
NM_001331927.1 | NP_001320611.1 | 23 - 24 | 3139 | 74 | ||
NM_100952.4 | NP_563877.1 | 25 - 26 | 3124 | 74 | ||
GenBank mRNA | AF312027.1 | - | 22 - 23 | 2830 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |