Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331926.1 | NP_001318975.1 | 33 - 34 | 4454 | 125 | |
NM_001331927.1 | NP_001320611.1 | 32 - 33 | 4303 | 125 | ||
NM_100952.4 | NP_563877.1 | 34 - 35 | 4288 | 125 | ||
GenBank mRNA | AF312027.1 | - | 31 - 32 | 3994 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |