Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331933.1 | NP_001320312.1 | 6 - 7 | 598 | 91 | |
NM_100962.4 | NP_172556.2 | 6 - 7 | 598 | 91 | ||
GenBank mRNA | AJ130878.1 | - | 6 - 7 | 533 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |