Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331933.1 | NP_001320312.1 | 9 - 10 | 811 | 113 | |
NM_100962.4 | NP_172556.2 | 9 - 10 | 811 | 113 | ||
GenBank mRNA | AJ130878.1 | - | 9 - 10 | 746 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |