Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RECQ4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100968.3 | NP_172562.2 | 6 - 7 | 1133 | 75 | |
GenBank mRNA | AY120761.1 | - | 6 - 7 | 1031 | 75 | |
BT010133.1 | - | 5 - 6 | 827 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |