Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scpl31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198028.1 | NP_001184957.1 | 5 - 6 | 750 | 121 | |
NM_100981.4 | NP_172575.2 | 5 - 6 | 955 | 121 | ||
GenBank mRNA | BT030381.1 | - | 5 - 6 | 701 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |