Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100998.4 | NP_172592.1 | 1 - 2 | 360 | 68 | |
GenBank mRNA | AY054249.1 | - | 1 - 2 | 248 | 68 | |
AY059781.1 | - | 1 - 2 | 266 | 68 | ||
AY133845.1 | - | 1 - 2 | 136 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |