Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345215.1 | NP_001331990.1 | 1 - 2 | 457 | 139 | |
NM_125057.4 | NP_200485.1 | 3 - 4 | 386 | 139 | ||
GenBank mRNA | BT025320.1 | - | 3 - 4 | 224 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |