Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BGAL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125070.3 | NP_200498.1 | 12 - 13 | 1802 | 144 | |
GenBank mRNA | AJ270300.1 | - | 12 - 13 | 1458 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |