Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAP1;3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125077.4 | NP_568844.1 | 3 - 4 | 353 | 68 | |
GenBank mRNA | AF370487.1 | - | 3 - 4 | 321 | 68 | |
AY064641.1 | - | 3 - 4 | 184 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |