Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345238.1 | NP_001331451.1 | 8 - 9 | 2155 | 72 | |
NM_125091.4 | NP_568848.1 | 8 - 9 | 2129 | 72 | ||
GenBank mRNA | AF056026.1 | - | 8 - 9 | 1898 | 72 | |
AF086906.1 | - | 8 - 9 | 1953 | 72 | ||
AF093241.1 | - | 8 - 9 | 1903 | 72 | ||
AK176358.1 | - | 8 - 9 | 1936 | 72 | ||
AY078965.1 | - | 8 - 9 | 1932 | 72 | ||
BT001152.1 | - | 8 - 9 | 1880 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |