Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAP28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125096.3 | NP_200524.1 | 3 - 4 | 798 | 85 | |
GenBank mRNA | AY136380.1 | - | 3 - 4 | 798 | 85 | |
AY882860.1 | - | 3 - 4 | 752 | 85 | ||
BT002110.1 | - | 3 - 4 | 752 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |