Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345261.1 | NP_001331405.1 | 6 - 7 | 777 | 80 | |
NM_125122.4 | NP_200549.2 | 7 - 8 | 1192 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |