Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VDAC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125130.3 | NP_200557.1 | 2 - 3 | 377 | 98 | |
GenBank mRNA | AY034928.1 | - | 2 - 3 | 231 | 98 | |
AY059112.1 | - | 1 - 2 | 45 | 98 | ||
AY086306.1 | - | 2 - 3 | 306 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |