Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345277.1 | NP_001318822.1 | 6 - 7 | 577 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |