Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBP23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125184.3 | NP_568873.1 | 4 - 5 | 894 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |