Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001085295.2 | NP_001078764.1 | 7 - 8 | 1088 | 88 | |
NM_001345284.1 | NP_001332019.1 | 6 - 7 | 1049 | 88 | ||
NM_001345285.1 | NP_001332020.1 | 6 - 7 | 1161 | 88 | ||
GenBank mRNA | DQ503426.1 | - | 6 - 7 | 957 | 88 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |