Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RR18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125193.4 | NP_200616.4 | 3 - 4 | 672 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |