Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125214.6 | NP_200637.1 | 3 - 4 | 950 | 69 | |
GenBank mRNA | AF123392.1 | - | 3 - 4 | 888 | 69 | |
AY070466.1 | - | 1363 | 69 | |||
BT020373.1 | - | 3 - 4 | 847 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |