Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125214.6 | NP_200637.1 | 4 - 5 | 1116 | 74 | |
GenBank mRNA | AF123392.1 | - | 4 - 5 | 1054 | 74 | |
AY070466.1 | - | 1529 | 74 | |||
BT020373.1 | - | 4 - 5 | 1013 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |