Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROC7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125258.5 | NP_200679.1 | 1 - 2 | 130 | 115 | |
GenBank mRNA | AF192490.1 | - | 1 - 2 | 88 | 115 | |
AY048227.1 | - | 1 - 2 | 79 | 115 | ||
AY086471.1 | - | 1 - 2 | 108 | 115 | ||
AY094017.1 | - | 1 - 2 | 65 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |