Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ftsh9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125277.4 | NP_568892.1 | 2 - 3 | 1154 | 80 | |
GenBank mRNA | AY059856.1 | - | 2 - 3 | 961 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |