Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RTFL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125343.3 | NP_200759.2 | 1 | 760 | 140 | |
GenBank mRNA | AY085568.1 | - | 1 | 441 | 140 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |