Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203644.1 | NP_001190573.1 | 11 - 12 | 1691 | 81 | |
NM_125397.3 | NP_200812.2 | 11 - 12 | 1557 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |