Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NRPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203644.1 | NP_001190573.1 | 18 - 19 | 2863 | 82 | |
NM_125397.3 | NP_200812.2 | 18 - 19 | 2708 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |