Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125462.4 | NP_200877.1 | 3 - 4 | 1224 | 70 | |
GenBank mRNA | AF188994.1 | - | 3 - 4 | 975 | 70 | |
AF233592.1 | - | 3 - 4 | 658 | 70 | ||
AK229564.1 | - | 3 - 4 | 1182 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |