Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125462.4 | NP_200877.1 | 8 - 9 | 1761 | 80 | |
GenBank mRNA | AF188994.1 | - | 8 - 9 | 1512 | 80 | |
AF233592.1 | - | 8 - 9 | 1195 | 80 | ||
AK229564.1 | - | 8 - 9 | 1719 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |