Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COBL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125488.5 | NP_200903.1 | 2 - 3 | 284 | 102 | |
GenBank mRNA | BT010831.1 | - | 2 - 3 | 217 | 102 | |
BT011296.1 | - | 2 - 3 | 138 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |