Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDA05 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203654.1 | NP_001190583.1 | 9 - 10 | 1520 | 98 | |
NM_125499.5 | NP_200914.2 | 9 - 10 | 1589 | 98 | ||
GenBank mRNA | AY090936.1 | - | 9 - 10 | 1508 | 98 | |
AY120784.1 | - | 9 - 10 | 1502 | 98 | ||
BT000073.1 | - | 9 - 10 | 1434 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |