Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNAP33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345457.1 | NP_001332102.1 | 2 - 3 | 998 | 83 | |
NM_125514.5 | NP_200929.1 | 2 - 3 | 829 | 83 | ||
GenBank mRNA | AY057627.1 | - | 2 - 3 | 744 | 83 | |
AY141994.1 | - | 1 - 2 | 474 | 83 | ||
X92419.1 | - | 715 | 83 | |||
X92420.1 | - | 660 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |