Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: PIF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037040.3 | NP_001032117.1 | 4 - 5 | 1451 | 122 | |
NM_001345463.1 | NP_001331793.1 | 5 - 6 | 1161 | 122 | ||
NM_125520.5 | NP_200935.2 | 5 - 6 | 1307 | 122 | ||
GenBank mRNA | AB493805.1 | - | 4 - 5 | 981 | 122 | |
AJ630484.1 | - | 4 - 5 | 717 | 122 | ||
AY568656.1 | - | 4 - 5 | 717 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |