Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACHT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125537.3 | NP_200952.1 | 3 - 4 | 752 | 109 | |
GenBank mRNA | AF144389.1 | - | 3 - 4 | 644 | 109 | |
AK229043.1 | - | 3 - 4 | 697 | 109 | ||
BT024467.1 | - | 3 - 4 | 539 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |