Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PFK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037043.1 | NP_001032120.1 | 5 - 6 | 764 | 105 | |
NM_125551.4 | NP_200966.2 | 5 - 6 | 814 | 105 | ||
GenBank mRNA | AY099694.1 | - | 5 - 6 | 762 | 105 | |
AY128873.1 | - | 5 - 6 | 705 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |