Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345493.1 | NP_001330984.1 | 1 - 2 | 389 | 71 | |
NM_125567.2 | NP_200981.1 | 1 - 2 | 367 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |