Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125573.4 | NP_200987.1 | 3 - 4 | 1538 | 105 | |
GenBank mRNA | AY059880.1 | - | 3 - 4 | 1391 | 105 | |
AY086864.1 | - | 3 - 4 | 1393 | 105 | ||
AY114669.1 | - | 3 - 4 | 1280 | 105 | ||
Z18242.1 | - | 1366 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |